siRNA Details
| Gene Symbol: Syngr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019166.2 | NP_062039.1 | 3 | 459 | ||
| XM_006241971.2 | XP_006242033.1 | 3 | 464 | |||
| GenBank mRNA | DN932005.1 | - | 3 | 421 | ||
| DV724604.1 | - | 2 | 265 | |||
| U39549.1 | - | 3 | 428 | |||

Syngr1
Genomic Map