siRNA Details
| Gene Symbol: Galns | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047851.1 | NP_001041316.1 | 2 | 194 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Galns| Gene Symbol: Galns | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047851.1 | NP_001041316.1 | 2 | 194 | ||