siRNA Details
| Gene Symbol: Galnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106196.1 | NP_001099666.1 | 12 | 1183 | ||
| XM_008772605.1 | XP_008770827.1 | 14 | 5140 | |||
| GenBank mRNA | BC168926.1 | - | 1394 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Galnt2| Gene Symbol: Galnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106196.1 | NP_001099666.1 | 12 | 1183 | ||
| XM_008772605.1 | XP_008770827.1 | 14 | 5140 | |||
| GenBank mRNA | BC168926.1 | - | 1394 | |||