siRNA Details
Gene Symbol: Galnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106196.1 | NP_001099666.1 | 5, 6 | 474 | ||
XM_008772605.1 | XP_008770827.1 | 6, 7 | 4338 | |||
GenBank mRNA | BC168926.1 | - | 592 |
