siRNA Details
| Gene Symbol: Epha3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031564.1 | NP_113752.1 | 4 | 882 | ||
| XM_006247976.2 | XP_006248038.1 | 4 | 966 | |||
| XM_006247977.2 | XP_006248039.1 | 4 | 2371 | |||
| XM_006247978.2 | XP_006248040.1 | 3 | 895 | |||
| XM_008768529.1 | XP_008766751.1 | 3 | 2005 | |||
| XM_008768530.1 | XP_008766752.1 | 3 | 769 | |||
| GenBank mRNA | U69278.1 | - | 4 | 882 | ||

Epha3
Genomic Map