siRNA Details
| Gene Symbol: RGD1311558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760533.1 | XP_008758755.1 | 13, 14 | 1516 | ||
| GenBank mRNA | BC085922.1 | - | 15, 16 | 1662 | ||
| EF055488.1 | - | 16, 17 | 1672 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1311558| Gene Symbol: RGD1311558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760533.1 | XP_008758755.1 | 13, 14 | 1516 | ||
| GenBank mRNA | BC085922.1 | - | 15, 16 | 1662 | ||
| EF055488.1 | - | 16, 17 | 1672 | |||