siRNA Details
Gene Symbol: Tubb4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080882.1 | NP_543158.1 | 4 | 677 | ||
GenBank mRNA | BC168216.1 | - | 4 | 682 | 7315263 | |
DY315612.1 | - | 1 | 238 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tubb4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080882.1 | NP_543158.1 | 4 | 677 | ||
GenBank mRNA | BC168216.1 | - | 4 | 682 | 7315263 | |
DY315612.1 | - | 1 | 238 |