siRNA Details
Gene Symbol: Mcoln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039005.1 | NP_001034094.1 | 1, 2 | 134 | ||
GenBank mRNA | BC099787.1 | - | 1, 2 | 134 | 7934506 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mcoln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039005.1 | NP_001034094.1 | 1, 2 | 134 | ||
GenBank mRNA | BC099787.1 | - | 1, 2 | 134 | 7934506 |