siRNA Details
| Gene Symbol: Lrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030827.1 | NP_110454.1 | 39 | 7468 | ||
| GenBank mRNA | L34049.1 | - | 39 | 7468 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrp2| Gene Symbol: Lrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030827.1 | NP_110454.1 | 39 | 7468 | ||
| GenBank mRNA | L34049.1 | - | 39 | 7468 | ||