siRNA Details
| Gene Symbol: Trip13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011930.1 | NP_001011930.1 | 2 | 322 | ||
| XM_008758681.1 | XP_008756903.1 | 2 | 324 | |||
| GenBank mRNA | BC083900.1 | - | 2 | 322 | 7133557 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trip13| Gene Symbol: Trip13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011930.1 | NP_001011930.1 | 2 | 322 | ||
| XM_008758681.1 | XP_008756903.1 | 2 | 324 | |||
| GenBank mRNA | BC083900.1 | - | 2 | 322 | 7133557 | |