siRNA Details
Gene Symbol: Ccdc170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758712.1 | XP_008756934.1 | 1816 | |||
XM_214760.8 | XP_214760.4 | 1817 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758712.1 | XP_008756934.1 | 1816 | |||
XM_214760.8 | XP_214760.4 | 1817 |