siRNA Details
| Gene Symbol: RGD1305235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017447.1 | NP_001017447.1 | 5 | 1350 | ||
| XM_006227833.2 | XP_006227895.1 | 4 | 1488 | |||
| GenBank mRNA | BC078920.1 | - | 5 | 1350 | 7096641 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1305235| Gene Symbol: RGD1305235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017447.1 | NP_001017447.1 | 5 | 1350 | ||
| XM_006227833.2 | XP_006227895.1 | 4 | 1488 | |||
| GenBank mRNA | BC078920.1 | - | 5 | 1350 | 7096641 | |