siRNA Details
| Gene Symbol: Nfib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031566.1 | NP_113754.1 | 9 | 1348 | ||
| XM_006238335.2 | XP_006238397.1 | 9 | 2013 | |||
| XM_006238336.2 | XP_006238398.1 | 9 | 2013 | |||
| XM_008763762.1 | XP_008761984.1 | 9 | 1755 | |||
| GenBank mRNA | AB012230.2 | - | 9 | 1348 | ||
| AB012231.2 | - | 9 | 1348 | |||

Nfib
Genomic Map