siRNA Details
| Gene Symbol: Sft2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008302.1 | NP_001008303.1 | 7, 8 | 506 | ||
| GenBank mRNA | BC085346.1 | - | 7, 8 | 506 | 7304630 | |
| FQ225074.1 | - | 6, 7 | 468 | |||
| FQ227930.1 | - | 7, 8 | 484 | |||

Sft2d1
Genomic Map