siRNA Details
| Gene Symbol: Thbs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169138.1 | NP_001162609.1 | 3 | 439 | ||
| GenBank mRNA | CB608927.1 | - | 3 | 424 | ||
| CB614734.1 | - | 3 | 416 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thbs2| Gene Symbol: Thbs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169138.1 | NP_001162609.1 | 3 | 439 | ||
| GenBank mRNA | CB608927.1 | - | 3 | 424 | ||
| CB614734.1 | - | 3 | 416 | |||