siRNA Details
| Gene Symbol: Lrp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106217.1 | NP_001099687.1 | 6, 7 | 1388 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrp11| Gene Symbol: Lrp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106217.1 | NP_001099687.1 | 6, 7 | 1388 | ||