siRNA Details
| Gene Symbol: Katna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004217.4 | NP_001004217.2 | 9 | 1262 | ||
| GenBank mRNA | AY621629.1 | - | 8 | 1066 | ||
| BC097929.1 | - | 9 | 1262 | 7444887 | ||
| BI302698.1 | - | 3 | 587 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Katna1| Gene Symbol: Katna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004217.4 | NP_001004217.2 | 9 | 1262 | ||
| GenBank mRNA | AY621629.1 | - | 8 | 1066 | ||
| BC097929.1 | - | 9 | 1262 | 7444887 | ||
| BI302698.1 | - | 3 | 587 | |||