siRNA Details
Gene Symbol: Stmn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024346.1 | NP_077322.1 | 4 | 395 | ||
GenBank mRNA | AY004290.1 | - | 4 | 395 | ||
FQ211987.1 | - | 3 | 247 | |||
FQ212062.1 | - | 3 | 256 | |||
FQ213831.1 | - | 3 | 250 | |||
FQ214184.1 | - | 3 | 252 |
