siRNA Details
| Gene Symbol: Stmn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024346.1 | NP_077322.1 | 4 | 455 | ||
| GenBank mRNA | AY004290.1 | - | 4 | 455 | ||
| FQ211987.1 | - | 3 | 307 | |||
| FQ212062.1 | - | 3 | 316 | |||
| FQ213831.1 | - | 3 | 310 | |||
| FQ214184.1 | - | 3 | 312 | |||

Stmn3
Genomic Map