siRNA Details
| Gene Symbol: Tnni3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017144.2 | NP_058840.1 | 6 | 443 | ||
| GenBank mRNA | DQ062462.1 | - | 4 | 339 | ||
| M57679.1 | - | 6 | 426 | |||
| M92074.1 | - | 5 | 393 | |||
| X58499.1 | - | 6 | 444 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnni3| Gene Symbol: Tnni3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017144.2 | NP_058840.1 | 6 | 443 | ||
| GenBank mRNA | DQ062462.1 | - | 4 | 339 | ||
| M57679.1 | - | 6 | 426 | |||
| M92074.1 | - | 5 | 393 | |||
| X58499.1 | - | 6 | 444 | |||