siRNA Details
| Gene Symbol: Tnni3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017144.2 | NP_058840.1 | 8 | 676 | ||
| GenBank mRNA | DQ062462.1 | - | 6 | 572 | ||
| M57679.1 | - | 8 | 659 | |||
| M92074.1 | - | 7 | 626 | |||
| X58499.1 | - | 8 | 677 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnni3| Gene Symbol: Tnni3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017144.2 | NP_058840.1 | 8 | 676 | ||
| GenBank mRNA | DQ062462.1 | - | 6 | 572 | ||
| M57679.1 | - | 8 | 659 | |||
| M92074.1 | - | 7 | 626 | |||
| X58499.1 | - | 8 | 677 | |||