siRNA Details
| Gene Symbol: Gltscr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106226.2 | NP_001099696.2 | 8 | 3161 | ||
| XM_006228343.2 | XP_006228405.1 | 10 | 3508 | |||
| XM_006228344.2 | XP_006228406.1 | 9 | 3442 | |||
| XM_006228345.2 | XP_006228407.1 | 9 | 3331 | |||

Gltscr1
Genomic Map