siRNA Details
| Gene Symbol: Mcpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172044.1 | NP_742041.1 | 4 | 483 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mcpt2| Gene Symbol: Mcpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172044.1 | NP_742041.1 | 4 | 483 | ||