siRNA Details
| Gene Symbol: Ethe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106234.1 | NP_001099704.1 | 2 | 235 | ||
| GenBank mRNA | BC158848.1 | - | 2 | 195 | 7444454 | |
| FQ209618.1 | - | 2 | 206 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ethe1| Gene Symbol: Ethe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106234.1 | NP_001099704.1 | 2 | 235 | ||
| GenBank mRNA | BC158848.1 | - | 2 | 195 | 7444454 | |
| FQ209618.1 | - | 2 | 206 | |||