siRNA Details
| Gene Symbol: Fbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025643.1 | NP_001020814.1 | 4 | 642 | ||
| GenBank mRNA | BC099198.1 | - | 4 | 642 | 7383920 | |
| FQ230160.1 | - | 2 | 195 | |||
| FQ231058.1 | - | 2 | 195 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbl| Gene Symbol: Fbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025643.1 | NP_001020814.1 | 4 | 642 | ||
| GenBank mRNA | BC099198.1 | - | 4 | 642 | 7383920 | |
| FQ230160.1 | - | 2 | 195 | |||
| FQ231058.1 | - | 2 | 195 | |||