siRNA Details
| Gene Symbol: Fbxo17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013064.2 | NP_001013082.2 | 5 | 770 | ||
| GenBank mRNA | BC079218.1 | - | 5 | 770 | 7131902 | |
| BE120593.1 | - | 3 | 436 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo17| Gene Symbol: Fbxo17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013064.2 | NP_001013082.2 | 5 | 770 | ||
| GenBank mRNA | BC079218.1 | - | 5 | 770 | 7131902 | |
| BE120593.1 | - | 3 | 436 | |||