siRNA Details
| Gene Symbol: Actc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019183.1 | NP_062056.1 | 5, 6 | 1006 | ||
| GenBank mRNA | BC127451.1 | - | 5, 6 | 881 | 7379877 | |
| CF109672.1 | - | 5, 6 | 860 | |||
| X80130.1 | - | 5, 6 | 892 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Actc1| Gene Symbol: Actc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019183.1 | NP_062056.1 | 5, 6 | 1006 | ||
| GenBank mRNA | BC127451.1 | - | 5, 6 | 881 | 7379877 | |
| CF109672.1 | - | 5, 6 | 860 | |||
| X80130.1 | - | 5, 6 | 892 | |||