siRNA Details
| Gene Symbol: Fam98c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198584.1 | NP_001185513.1 | 4 | 525 | ||
| XM_006228717.2 | XP_006228779.1 | 4 | 555 | |||
| GenBank mRNA | BC079080.1 | - | 3 | 563 | 7122702 | |
| CO564588.1 | - | 2 | 302 | |||
| CO570848.1 | - | 4 | 525 | |||
| DV214726.1 | - | 4 | 550 | |||

Fam98c
Genomic Map