siRNA Details
| Gene Symbol: Psmd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100831.1 | NP_001094301.1 | 1 | 365 | ||
| GenBank mRNA | BC105894.1 | - | 1 | 130 | ||
| CD371338.1 | - | 1 | 375 | |||
| FQ213223.1 | - | 1 | 165 | |||
| FQ214314.1 | - | 1 | 162 | |||
| FQ223219.1 | - | 1 | 208 | |||
| FQ233693.1 | - | 1 | 196 | |||

Psmd8
Genomic Map