siRNA Details
| Gene Symbol: Sipa1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013066.1 | NP_001013084.1 | 10 | 1859 | ||
| XM_008759130.1 | XP_008757352.1 | 20 | 5286 | |||
| XM_008759131.1 | XP_008757353.1 | 20 | 5283 | |||
| GenBank mRNA | BC091303.1 | - | 10 | 1859 | 7309905 | |

Sipa1l3
Genomic Map