siRNA Details
Gene Symbol: Lasp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032613.2 | NP_116002.1 | 7 | 782 | ||
XM_006247288.2 | XP_006247350.1 | 7 | 714 | |||
GenBank mRNA | AF242187.1 | - | 7 | 696 | ||
BC099791.1 | - | 7 | 782 | 7934892 |
