siRNA Details
Gene Symbol: Tmem147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038494.1 | NP_001033583.1 | 2 | 239 | ||
GenBank mRNA | BC111082.1 | - | 2 | 239 | 7455198 | |
FQ234356.1 | - | 2 | 254 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038494.1 | NP_001033583.1 | 2 | 239 | ||
GenBank mRNA | BC111082.1 | - | 2 | 239 | 7455198 | |
FQ234356.1 | - | 2 | 254 |