siRNA Details
| Gene Symbol: Pdcd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106247.1 | NP_001099717.1 | 4 | 343 | ||
| XM_006228884.2 | XP_006228946.1 | 4 | 211 | |||
| GenBank mRNA | FQ222074.1 | - | 4 | 258 | ||
| Gene Symbol: LOC100912106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748838.3 | XP_003748886.1 | 4 | 347 | ||
| XM_006228897.2 | XP_006228959.1 | 4 | 211 | |||

Pdcd5
Genomic Map