siRNA Details
| Gene Symbol: Etfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004220.1 | NP_001004220.1 | 1, 2 | 84 | ||
| GenBank mRNA | BC079351.1 | - | 1, 2 | 84 | 7108112 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Etfb| Gene Symbol: Etfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004220.1 | NP_001004220.1 | 1, 2 | 84 | ||
| GenBank mRNA | BC079351.1 | - | 1, 2 | 84 | 7108112 | |