siRNA Details
Gene Symbol: Rps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017151.2 | NP_058847.1 | 4 | 371 | ||
GenBank mRNA | AA892895.1 | - | 1 | 116 | ||
AI603168.1 | - | 1 | 134 | |||
AY390387.1 | - | 3 | 299 | |||
FQ211784.1 | - | 4 | 371 | |||
FQ217117.1 | - | 4 | 372 | |||
FQ217388.1 | - | 4 | 371 | |||
FQ217914.1 | - | 4 | 371 | |||
FQ220999.1 | - | 4 | 371 | |||
FQ221086.1 | - | 4 | 372 | |||
FQ221189.1 | - | 4 | 371 | |||
FQ221217.1 | - | 4 | 372 | |||
FQ221800.1 | - | 4 | 371 | |||
FQ222134.1 | - | 4 | 359 | |||
FQ222137.1 | - | 4 | 372 | |||
FQ222200.1 | - | 4 | 371 | |||
FQ222233.1 | - | 4 | 370 | |||
FQ222539.1 | - | 4 | 371 | |||
FQ222635.1 | - | 4 | 374 | |||
FQ222928.1 | - | 4 | 371 | |||
FQ223014.1 | - | 3 | 373 | |||
FQ223247.1 | - | 4 | 371 | |||
FQ223311.1 | - | 4 | 372 | |||
FQ223478.1 | - | 4 | 374 | |||
FQ223811.1 | - | 4 | 371 | |||
FQ223860.1 | - | 4 | 367 | |||
FQ224530.1 | - | 4 | 371 | |||
FQ228367.1 | - | 4 | 379 | |||
FQ228405.1 | - | 4 | 374 | |||
M19393.1 | - | 4 | 371 |
