siRNA Details
Gene Symbol: LOC102553035 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229178.2 | XP_006229240.2 | 2, 3 | 262 | ||
XM_008759447.1 | XP_008757669.1 | 3, 4 | 242 |
Gene Symbol: Klk15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728759.4 | XP_002728805.2 | 3, 4 | 407 |
