siRNA Details
Gene Symbol: Josd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106256.2 | NP_001099726.2 | 4 | 443 | ||
XM_006228996.2 | XP_006229058.1 | 5 | 725 | |||
XM_006228997.2 | XP_006229059.1 | 5 | 722 | |||
XM_006228998.2 | XP_006229060.1 | 4 | 664 | |||
GenBank mRNA | CK842299.1 | - | 2 | 422 | ||
DV214517.1 | - | 4 | 443 |
