siRNA Details
Gene Symbol: Emc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004221.1 | NP_001004221.1 | 4 | 454 | ||
XM_006229000.2 | XP_006229062.1 | 4 | 470 | |||
XM_006229001.2 | XP_006229063.1 | 4 | 470 | |||
GenBank mRNA | BC079041.1 | - | 4 | 454 | 7114835 |
