siRNA Details
| Gene Symbol: Ruvbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025405.2 | NP_001020576.2 | 14 | 1509 | ||
| XM_006229029.1 | XP_006229091.1 | 14 | 1627 | |||
| GenBank mRNA | BC082022.1 | - | 12 | 1444 | ||
| BC098042.1 | - | 13 | 1279 | 7457010 | ||

Ruvbl2
Genomic Map