siRNA Details
| Gene Symbol: Ruvbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025405.2 | NP_001020576.2 | 14, 15 | 1594 | ||
| GenBank mRNA | BC082022.1 | - | 12, 13 | 1529 | ||
| BC098042.1 | - | 13, 14 | 1364 | 7457010 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ruvbl2| Gene Symbol: Ruvbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025405.2 | NP_001020576.2 | 14, 15 | 1594 | ||
| GenBank mRNA | BC082022.1 | - | 12, 13 | 1529 | ||
| BC098042.1 | - | 13, 14 | 1364 | 7457010 | ||