siRNA Details
| Gene Symbol: Adamts17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229360.2 | XP_006229422.1 | 2 | 587 | ||
| XM_218753.8 | XP_218753.6 | 2 | 586 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adamts17| Gene Symbol: Adamts17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229360.2 | XP_006229422.1 | 2 | 587 | ||
| XM_218753.8 | XP_218753.6 | 2 | 586 | |||