siRNA Details
| Gene Symbol: Klhl25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039006.1 | NP_001034095.1 | 2 | 1841 | ||
| XM_006229379.2 | XP_006229441.1 | 2 | 2154 | |||
| XM_006229380.2 | XP_006229442.1 | 2 | 6192 | |||
| XM_006229381.2 | XP_006229443.1 | 2 | 5247 | |||
| XM_008759505.1 | XP_008757727.1 | 2 | 1884 | |||
| GenBank mRNA | BC099111.1 | - | 2 | 1841 | 7461562 | |
| FQ230538.1 | - | 2 | 1849 | |||

Klhl25
Genomic Map