siRNA Details
Gene Symbol: Zfp710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759507.1 | XP_008757729.1 | 5 | 3765 | ||
XM_008759508.1 | XP_008757730.1 | 5 | 3570 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759507.1 | XP_008757729.1 | 5 | 3765 | ||
XM_008759508.1 | XP_008757730.1 | 5 | 3570 |