siRNA Details
Gene Symbol: Rlbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106274.1 | NP_001099744.1 | 4, 5 | 701 | ||
XM_006229396.2 | XP_006229458.1 | 4, 5 | 617 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rlbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106274.1 | NP_001099744.1 | 4, 5 | 701 | ||
XM_006229396.2 | XP_006229458.1 | 4, 5 | 617 |