siRNA Details
| Gene Symbol: Rlbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106274.1 | NP_001099744.1 | 6 | 948 | ||
| XM_006229396.2 | XP_006229458.1 | 6 | 864 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rlbp1| Gene Symbol: Rlbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106274.1 | NP_001099744.1 | 6 | 948 | ||
| XM_006229396.2 | XP_006229458.1 | 6 | 864 | |||