siRNA Details
| Gene Symbol: Rps6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017160.1 | NP_058856.1 | 143 | |||
| GenBank mRNA | BC058149.1 | - | 173 | 6918741 | ||
| M29358.1 | - | 143 | ||||
| Gene Symbol: LOC100911372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749971.3 | XP_003750019.1 | 2, 3 | 228 | ||

Rps6
Genomic Map