siRNA Details
Gene Symbol: Rps6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017160.1 | NP_058856.1 | 409 | |||
GenBank mRNA | BC058149.1 | - | 439 | 6918741 | ||
FQ210697.1 | - | 41 | ||||
FQ211324.1 | - | 41 | ||||
FQ212249.1 | - | 41 | ||||
FQ217720.1 | - | 41 | ||||
FQ218111.1 | - | 41 | ||||
FQ221402.1 | - | 41 | ||||
FQ222806.1 | - | 41 | ||||
FQ229168.1 | - | 41 | ||||
FQ230362.1 | - | 41 | ||||
FQ231606.1 | - | 41 | ||||
FQ235053.1 | - | 41 | ||||
M29358.1 | - | 409 |
Gene Symbol: LOC100911372 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749971.3 | XP_003750019.1 | 4 | 494 |
