siRNA Details
Gene Symbol: Fam103a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127451.1 | NP_001120923.1 | 3 | 461 | ||
GenBank mRNA | AA900118.1 | - | 1 | 348 | ||
BC089896.1 | - | 2 | 1110 | 7381229 | ||
CF977463.1 | - | 3 | 481 | |||
DV717487.1 | - | 2 | 314 |
