siRNA Details
| Gene Symbol: RGD1305713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024749.1 | NP_001019920.1 | 3 | 326 | ||
| XM_006229429.2 | XP_006229491.1 | 3 | 2031 | |||
| GenBank mRNA | BC094529.1 | - | 3 | 326 | 7320095 | |
| Gene Symbol: LOC100363116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750235.3 | XP_003750283.1 | 1 | 595 | ||

RGD1305713
Genomic Map