siRNA Details
| Gene Symbol: Btbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011932.1 | NP_001011932.1 | 2 | 506 | ||
| GenBank mRNA | BC085850.1 | - | 2 | 506 | 7189751 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Btbd1| Gene Symbol: Btbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011932.1 | NP_001011932.1 | 2 | 506 | ||
| GenBank mRNA | BC085850.1 | - | 2 | 506 | 7189751 | |